Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612738 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC9C1 - solute carrier family 9 member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320531.1 | Intron | NP_001307460.1 | ||||
NM_183061.2 | Intron | NP_898884.1 | ||||
XM_006713603.2 | Intron | XP_006713666.1 | ||||
XM_011512718.1 | Intron | XP_011511020.1 | ||||
XM_011512719.1 | Intron | XP_011511021.1 | ||||
XM_011512720.1 | Intron | XP_011511022.1 | ||||
XM_011512721.1 | Intron | XP_011511023.1 | ||||
XM_011512724.1 | Intron | XP_011511026.1 | ||||
XM_011512725.2 | Intron | XP_011511027.1 | ||||
XM_011512726.2 | Intron | XP_011511028.1 | ||||
XM_011512727.2 | Intron | XP_011511029.1 | ||||
XM_017006246.1 | Intron | XP_016861735.1 | ||||
XM_017006247.1 | Intron | XP_016861736.1 | ||||
XM_017006248.1 | Intron | XP_016861737.1 | ||||
XM_017006249.1 | Intron | XP_016861738.1 | ||||
XM_017006250.1 | Intron | XP_016861739.1 |
Set Membership: |
HapMap |