Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608467 | ||||||||||||||||||||
Literature Links: |
STON2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STON2 - stonin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256430.1 | 6208 | Intron | NP_001243359.1 | |||
NM_033104.3 | 6208 | UTR 3 | NP_149095.2 | |||
XM_006720279.1 | 6208 | UTR 3 | XP_006720342.1 | |||
XM_011537240.1 | 6208 | Intron | XP_011535542.1 | |||
XM_011537241.1 | 6208 | Intron | XP_011535543.1 | |||
XM_011537242.1 | 6208 | UTR 3 | XP_011535544.1 |