Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605018 | ||||||||||||||||||||
Literature Links: |
CYLD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYLD - CYLD lysine 63 deubiquitinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042355.1 | Intron | NP_001035814.1 | ||||
NM_001042412.1 | Intron | NP_001035877.1 | ||||
NM_015247.2 | Intron | NP_056062.1 | ||||
XM_005255812.2 | Intron | XP_005255869.1 | ||||
XM_006721149.1 | Intron | XP_006721212.1 | ||||
XM_011522907.2 | Intron | XP_011521209.1 | ||||
XM_011522908.1 | Intron | XP_011521210.1 | ||||
XM_011522911.2 | Intron | XP_011521213.1 | ||||
XM_017022977.1 | Intron | XP_016878466.1 | ||||
XM_017022978.1 | Intron | XP_016878467.1 | ||||
XM_017022979.1 | Intron | XP_016878468.1 | ||||
XM_017022980.1 | Intron | XP_016878469.1 | ||||
XM_017022981.1 | Intron | XP_016878470.1 |