Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604978 MIM: 615857 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUDT21 - nudix hydrolase 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OGFOD1 - 2-oxoglutarate and iron dependent oxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324357.1 | Intron | NP_001311286.1 | ||||
NM_001324358.1 | Intron | NP_001311287.1 | ||||
NM_001324359.1 | Intron | NP_001311288.1 | ||||
NM_001324360.1 | Intron | NP_001311289.1 | ||||
NM_001324361.1 | Intron | NP_001311290.1 | ||||
NM_001324362.1 | Intron | NP_001311291.1 | ||||
NM_001324363.1 | Intron | NP_001311292.1 | ||||
NM_018233.3 | Intron | NP_060703.3 |
Set Membership: |
HapMap |