Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605405 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP6NL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP6NL - USP6 N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080491.3 | 7480 | UTR 3 | NP_001073960.1 | |||
NM_014688.3 | 7480 | UTR 3 | NP_055503.1 | |||
XM_006717542.3 | 7480 | UTR 3 | XP_006717605.1 | |||
XM_011519762.2 | 7480 | UTR 3 | XP_011518064.1 | |||
XM_011519763.2 | 7480 | UTR 3 | XP_011518065.1 | |||
XM_017016971.1 | 7480 | UTR 3 | XP_016872460.1 | |||
XM_017016972.1 | 7480 | UTR 3 | XP_016872461.1 | |||
XM_017016973.1 | 7480 | UTR 3 | XP_016872462.1 |
Set Membership: |
HapMap |