Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAACCTGCCTAAGTTCACCTCCT[G/T]GTGGCCACTGTTCTTCTATTACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607591 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf44-SGK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C8orf44-SGK3 - C8orf44-SGK3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204173.1 | Intron | NP_001191102.1 |
SGK3 - serum/glucocorticoid regulated kinase family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033578.2 | Intron | NP_001028750.1 | ||||
NM_013257.4 | Intron | NP_037389.4 | ||||
NM_170709.2 | Intron | NP_733827.2 |
Set Membership: |
HapMap |