Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600904 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASTN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASTN1 - astrotactin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286164.1 | 4920 | Intron | NP_001273093.1 | |||
NM_004319.2 | 4920 | UTR 3 | NP_004310.1 | |||
NM_207108.2 | 4920 | Intron | NP_996991.1 | |||
XM_017001340.1 | 4920 | UTR 3 | XP_016856829.1 | |||
XM_017001341.1 | 4920 | Intron | XP_016856830.1 |
Set Membership: |
HapMap |