Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCSTAMP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCSTAMP - dendrocyte expressed seven transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257317.1 | Intron | NP_001244246.1 | ||||
NM_030788.3 | Intron | NP_110415.1 | ||||
XM_011517321.1 | Intron | XP_011515623.1 | ||||
XM_011517324.1 | Intron | XP_011515626.1 | ||||
XM_017013878.1 | Intron | XP_016869367.1 | ||||
XM_017013879.1 | Intron | XP_016869368.1 | ||||
XM_017013880.1 | Intron | XP_016869369.1 |