Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605401 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQGAP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IQGAP2 - IQ motif containing GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285460.1 | Intron | NP_001272389.1 | ||||
NM_001285461.1 | Intron | NP_001272390.1 | ||||
NM_001285462.1 | Intron | NP_001272391.1 | ||||
NM_006633.3 | Intron | NP_006624.2 | ||||
XM_005248410.2 | Intron | XP_005248467.1 | ||||
XM_005248414.1 | Intron | XP_005248471.1 | ||||
XM_011543108.1 | Intron | XP_011541410.1 | ||||
XM_017008960.1 | Intron | XP_016864449.1 |
Set Membership: |
HapMap |