Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKA - diacylglycerol kinase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001345.4 | 210 | Silent Mutation | GTC,GTG | V,V 43 | NP_001336.2 | |
NM_201444.2 | 210 | Silent Mutation | GTC,GTG | V,V 43 | NP_958852.1 | |
NM_201445.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | NP_958853.1 | |
NM_201554.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | NP_963848.1 | |
XM_005268688.2 | 210 | Silent Mutation | GTC,GTG | V,V 81 | XP_005268745.1 | |
XM_005268689.2 | 210 | UTR 5 | XP_005268746.1 | |||
XM_005268690.2 | 210 | UTR 5 | XP_005268747.1 | |||
XM_011537991.2 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_011536293.1 | |
XM_011537993.2 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_011536295.1 | |
XM_011537995.2 | 210 | UTR 5 | XP_011536297.1 | |||
XM_017018900.1 | 210 | Silent Mutation | GTC,GTG | V,V 81 | XP_016874389.1 | |
XM_017018901.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874390.1 | |
XM_017018902.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874391.1 | |
XM_017018903.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874392.1 | |
XM_017018904.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874393.1 | |
XM_017018905.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874394.1 | |
XM_017018906.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874395.1 | |
XM_017018907.1 | 210 | Silent Mutation | GTC,GTG | V,V 43 | XP_016874396.1 | |
XM_017018908.1 | 210 | Silent Mutation | GTC,GTG | V,V 81 | XP_016874397.1 | |
XM_017018909.1 | 210 | UTR 5 | XP_016874398.1 |
PYM1 - PYM homolog 1, exon junction complex associated factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |