Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608368 MIM: 606932 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD9B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAD9B - RAD9 checkpoint clamp component B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286531.1 | Intron | NP_001273460.1 | ||||
NM_001286532.1 | Intron | NP_001273461.1 | ||||
NM_001286533.1 | Intron | NP_001273462.1 | ||||
NM_001286534.1 | Intron | NP_001273463.1 | ||||
NM_001286535.1 | Intron | NP_001273464.1 | ||||
NM_001286536.1 | Intron | NP_001273465.1 | ||||
NM_152442.3 | Intron | NP_689655.3 | ||||
XM_005253849.4 | Intron | XP_005253906.1 | ||||
XM_011537971.2 | Intron | XP_011536273.1 | ||||
XM_011537972.2 | Intron | XP_011536274.1 | ||||
XM_011537974.2 | Intron | XP_011536276.1 | ||||
XM_017018877.1 | Intron | XP_016874366.1 | ||||
XM_017018878.1 | Intron | XP_016874367.1 | ||||
XM_017018879.1 | Intron | XP_016874368.1 | ||||
XM_017018880.1 | Intron | XP_016874369.1 | ||||
XM_017018881.1 | Intron | XP_016874370.1 | ||||
XM_017018882.1 | Intron | XP_016874371.1 |
VPS29 - VPS29, retromer complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282150.1 | Intron | NP_001269079.1 | ||||
NM_001282151.1 | Intron | NP_001269080.1 | ||||
NM_016226.4 | Intron | NP_057310.1 | ||||
NM_057180.2 | Intron | NP_476528.1 | ||||
XM_006719459.3 | Intron | XP_006719522.1 | ||||
XM_006719460.3 | Intron | XP_006719523.1 | ||||
XM_011538484.2 | Intron | XP_011536786.1 | ||||
XM_017019457.1 | Intron | XP_016874946.1 | ||||
XM_017019458.1 | Intron | XP_016874947.1 | ||||
XM_017019459.1 | Intron | XP_016874948.1 |
Set Membership: |
HapMap |