Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137167 MIM: 601468 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGCX PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GGCX - gamma-glutamyl carboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000821.6 | 3231 | UTR 3 | NP_000812.2 | |||
NM_001142269.3 | 3231 | UTR 3 | NP_001135741.1 | |||
NM_001311312.1 | 3231 | Intron | NP_001298241.1 | |||
XM_005264259.4 | 3231 | UTR 3 | XP_005264316.1 | |||
XM_011532764.2 | 3231 | UTR 3 | XP_011531066.1 | |||
XM_011532765.2 | 3231 | UTR 3 | XP_011531067.1 | |||
XM_017003803.1 | 3231 | UTR 3 | XP_016859292.1 |
MAT2A - methionine adenosyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |