Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613555 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGUOK-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGUOK-AS1 - DGUOK antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TET3 - tet methylcytosine dioxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287491.1 | 1280 | Intron | NP_001274420.1 | |||
XM_005264187.3 | 1280 | Intron | XP_005264244.1 | |||
XM_011532682.2 | 1280 | Intron | XP_011530984.1 | |||
XM_011532683.2 | 1280 | Intron | XP_011530985.1 | |||
XM_011532684.2 | 1280 | UTR 5 | XP_011530986.1 | |||
XM_011532685.2 | 1280 | Intron | XP_011530987.1 | |||
XM_011532686.2 | 1280 | Intron | XP_011530988.1 | |||
XM_011532687.2 | 1280 | Intron | XP_011530989.1 | |||
XM_011532688.2 | 1280 | Intron | XP_011530990.1 | |||
XM_011532689.2 | 1280 | Intron | XP_011530991.1 | |||
XM_011532690.1 | 1280 | Intron | XP_011530992.1 | |||
XM_017003566.1 | 1280 | UTR 5 | XP_016859055.1 | |||
XM_017003567.1 | 1280 | Intron | XP_016859056.1 |