Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613503 MIM: 615161 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-DQA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HLA-DQA2 - major histocompatibility complex, class II, DQ alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HLA-DQB2 - major histocompatibility complex, class II, DQ beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198858.1 | 850 | UTR 3 | NP_001185787.1 | |||
NM_001300790.1 | 850 | UTR 3 | NP_001287719.1 | |||
XM_005249051.4 | 850 | Intron | XP_005249108.1 | |||
XM_011514560.2 | 850 | UTR 3 | XP_011512862.1 | |||
XM_011514561.2 | 850 | UTR 3 | XP_011512863.1 |
MIR3135B - microRNA 3135b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |