Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTCTCAGGAAGGGTGTCTGTCT[C/G]ATGTTCCATCACCAACTTGTTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147267 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITPR3 - inositol 1,4,5-trisphosphate receptor type 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002224.3 | Intron | NP_002215.2 | ||||
XM_011514577.2 | Intron | XP_011512879.1 | ||||
XM_017010832.1 | Intron | XP_016866321.1 |
LOC101929188 - uncharacterized LOC101929188 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |