Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610514 MIM: 611399 | ||||||||||||||||||||
Literature Links: |
JADE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JADE1 - jade family PHD finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCLT1 - sodium channel and clathrin linker 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300897.1 | Intron | NP_001287826.1 | ||||
NM_001300898.1 | Intron | NP_001287827.1 | ||||
NM_144643.3 | Intron | NP_653244.2 | ||||
XM_017007716.1 | Intron | XP_016863205.1 | ||||
XM_017007717.1 | Intron | XP_016863206.1 | ||||
XM_017007718.1 | Intron | XP_016863207.1 | ||||
XM_017007719.1 | Intron | XP_016863208.1 | ||||
XM_017007720.1 | Intron | XP_016863209.1 | ||||
XM_017007721.1 | Intron | XP_016863210.1 |