Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615790 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHDC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AHDC1 - AT-hook DNA binding motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029882.3 | Intron | NP_001025053.1 | ||||
XM_005245848.3 | Intron | XP_005245905.1 | ||||
XM_005245849.3 | Intron | XP_005245906.1 | ||||
XM_005245850.3 | Intron | XP_005245907.1 | ||||
XM_005245851.3 | Intron | XP_005245908.1 | ||||
XM_005245852.3 | Intron | XP_005245909.1 | ||||
XM_011541255.1 | Intron | XP_011539557.1 | ||||
XM_011541256.2 | Intron | XP_011539558.1 | ||||
XM_011541257.2 | Intron | XP_011539559.1 |
Set Membership: |
HapMap |