Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACATGTTTCTAGGCAAAATACT[C/T]ACCTGCCATCCCTATTCATGTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609101 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO30 - F-box protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032145.4 | Intron | NP_115521.3 | ||||
XM_005267159.3 | Intron | XP_005267216.1 | ||||
XM_011536177.2 | Intron | XP_011534479.1 | ||||
XM_017011353.1 | Intron | XP_016866842.1 |
LOC100507557 - uncharacterized LOC100507557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |