Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604613 | ||||||||||||||||||||
Literature Links: |
TBX18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBX18 - T-box 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080508.2 | 1226 | Intron | NP_001073977.1 | |||
XM_006715602.3 | 1226 | Intron | XP_006715665.2 | |||
XM_011536247.2 | 1226 | Intron | XP_011534549.2 | |||
XM_011536248.2 | 1226 | Intron | XP_011534550.2 | |||
XM_011536249.2 | 1226 | Intron | XP_011534551.2 | |||
XM_017011472.1 | 1226 | Intron | XP_016866961.1 | |||
XM_017011473.1 | 1226 | UTR 3 | XP_016866962.1 |