Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113710 MIM: 605511 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TFF1 - trefoil factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS3 - transmembrane protease, serine 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256317.1 | Intron | NP_001243246.1 | ||||
NM_024022.2 | Intron | NP_076927.1 | ||||
NM_032404.2 | Intron | NP_115780.1 | ||||
NM_032405.1 | Intron | NP_115781.1 |
Set Membership: |
HapMap |