Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609525 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
METTL8 - methyltransferase like 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321154.1 | 8922 | UTR 3 | NP_001308083.1 | |||
NM_001321155.1 | 8922 | UTR 3 | NP_001308084.1 | |||
NM_001321156.1 | 8922 | UTR 3 | NP_001308085.1 | |||
NM_001321157.1 | 8922 | UTR 3 | NP_001308086.1 | |||
NM_001321158.1 | 8922 | UTR 3 | NP_001308087.1 | |||
NM_001321159.1 | 8922 | UTR 3 | NP_001308088.1 | |||
NM_001321160.1 | 8922 | UTR 3 | NP_001308089.1 | |||
NM_001321161.1 | 8922 | UTR 3 | NP_001308090.1 | |||
NM_001321162.1 | 8922 | UTR 3 | NP_001308091.1 | |||
NM_024770.4 | 8922 | UTR 3 | NP_079046.2 | |||
XM_011511875.2 | 8922 | Intron | XP_011510177.1 | |||
XM_017004984.1 | 8922 | Intron | XP_016860473.1 | |||
XM_017004985.1 | 8922 | Intron | XP_016860474.1 | |||
XM_017004986.1 | 8922 | Intron | XP_016860475.1 | |||
XM_017004987.1 | 8922 | Intron | XP_016860476.1 | |||
XM_017004988.1 | 8922 | Intron | XP_016860477.1 | |||
XM_017004989.1 | 8922 | Intron | XP_016860478.1 |
Set Membership: |
HapMap |