Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611917 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KDM8 - lysine demethylase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145348.1 | Intron | NP_001138820.1 | ||||
NM_024773.2 | Intron | NP_079049.2 | ||||
XM_017023676.1 | Intron | XP_016879165.1 | ||||
XM_017023677.1 | Intron | XP_016879166.1 | ||||
XM_017023678.1 | Intron | XP_016879167.1 | ||||
XM_017023679.1 | Intron | XP_016879168.1 | ||||
XM_017023680.1 | Intron | XP_016879169.1 |