Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD18A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD18A - ankyrin repeat domain 18A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147195.2 | Intron | NP_671728.2 | ||||
XM_006716749.3 | Intron | XP_006716812.1 | ||||
XM_011517836.2 | Intron | XP_011516138.1 | ||||
XM_011517837.2 | Intron | XP_011516139.1 | ||||
XM_011517839.2 | Intron | XP_011516141.1 | ||||
XM_017014576.1 | Intron | XP_016870065.1 | ||||
XM_017014577.1 | Intron | XP_016870066.1 |
FAM201A - family with sequence similarity 201 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |