Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZCCHC7 - zinc finger CCHC-type containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289119.1 | Intron | NP_001276048.1 | ||||
NM_001289120.1 | Intron | NP_001276049.1 | ||||
NM_001289121.1 | Intron | NP_001276050.1 | ||||
NM_032226.2 | Intron | NP_115602.2 | ||||
XM_005251608.4 | Intron | XP_005251665.1 | ||||
XM_005251612.2 | Intron | XP_005251669.1 | ||||
XM_011518050.1 | Intron | XP_011516352.1 | ||||
XM_011518051.2 | Intron | XP_011516353.1 | ||||
XM_017015197.1 | Intron | XP_016870686.1 | ||||
XM_017015198.1 | Intron | XP_016870687.1 | ||||
XM_017015199.1 | Intron | XP_016870688.1 | ||||
XM_017015200.1 | Intron | XP_016870689.1 | ||||
XM_017015201.1 | Intron | XP_016870690.1 |