Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605227 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RECK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RECK - reversion inducing cysteine rich protein with kazal motifs | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316345.1 | Intron | NP_001303274.1 | ||||
NM_001316346.1 | Intron | NP_001303275.1 | ||||
NM_001316347.1 | Intron | NP_001303276.1 | ||||
NM_001316348.1 | Intron | NP_001303277.1 | ||||
NM_021111.2 | Intron | NP_066934.1 | ||||
XM_017015207.1 | Intron | XP_016870696.1 | ||||
XM_017015208.1 | Intron | XP_016870697.1 | ||||
XM_017015209.1 | Intron | XP_016870698.1 | ||||
XM_017015210.1 | Intron | XP_016870699.1 | ||||
XM_017015211.1 | Intron | XP_016870700.1 |
Set Membership: |
HapMap |