Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601274 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTGR1 - prostaglandin reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146108.1 | Intron | NP_001139580.1 | ||||
NM_001146109.1 | Intron | NP_001139581.1 | ||||
NM_012212.3 | Intron | NP_036344.2 | ||||
XM_011518394.2 | Intron | XP_011516696.1 | ||||
XM_011518395.2 | Intron | XP_011516697.1 | ||||
XM_017014485.1 | Intron | XP_016869974.1 |
ZNF483 - zinc finger protein 483 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007169.3 | Intron | NP_001007170.1 | ||||
NM_133464.3 | Intron | NP_597721.2 | ||||
XM_011518300.2 | Intron | XP_011516602.1 | ||||
XM_017014337.1 | Intron | XP_016869826.1 | ||||
XM_017014338.1 | Intron | XP_016869827.1 | ||||
XM_017014339.1 | Intron | XP_016869828.1 |
Set Membership: |
HapMap |