Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610054 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf66 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C5orf66 - chromosome 5 open reading frame 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277348.1 | Intron | NP_001264277.1 |
H2AFY - H2A histone family member Y | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040158.1 | Intron | NP_001035248.1 | ||||
NM_004893.2 | Intron | NP_004884.1 | ||||
NM_138609.2 | Intron | NP_613075.1 | ||||
NM_138610.2 | Intron | NP_613258.2 | ||||
XM_005272132.1 | Intron | XP_005272189.1 | ||||
XM_005272134.3 | Intron | XP_005272191.1 | ||||
XM_005272135.3 | Intron | XP_005272192.1 | ||||
XM_011543729.2 | Intron | XP_011542031.1 | ||||
XM_011543730.2 | Intron | XP_011542032.1 | ||||
XM_011543731.1 | Intron | XP_011542033.1 | ||||
XM_011543732.1 | Intron | XP_011542034.1 | ||||
XM_011543733.2 | Intron | XP_011542035.1 | ||||
XM_011543734.2 | Intron | XP_011542036.1 | ||||
XM_011543735.2 | Intron | XP_011542037.1 | ||||
XM_017010067.1 | Intron | XP_016865556.1 | ||||
XM_017010068.1 | Intron | XP_016865557.1 | ||||
XM_017010069.1 | Intron | XP_016865558.1 |
Set Membership: |
HapMap |