Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL28 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLHL28 - kelch like family member 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308112.1 | 3968 | UTR 3 | NP_001295041.1 | |||
NM_017658.4 | 3968 | UTR 3 | NP_060128.2 | |||
XM_005267770.3 | 3968 | UTR 3 | XP_005267827.1 | |||
XM_006720174.3 | 3968 | Intron | XP_006720237.1 | |||
XM_011536847.2 | 3968 | UTR 3 | XP_011535149.1 | |||
XM_011536849.2 | 3968 | UTR 3 | XP_011535151.1 | |||
XM_017021378.1 | 3968 | Intron | XP_016876867.1 |
Set Membership: |
HapMap |