Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600754 MIM: 611856 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MMP14 - matrix metallopeptidase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL52 - mitochondrial ribosomal protein L52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318870.1 | Intron | NP_001305799.1 | ||||
NM_178336.2 | Intron | NP_848026.1 | ||||
NM_180982.2 | Intron | NP_851313.1 | ||||
NM_181304.2 | Intron | NP_851821.1 | ||||
NM_181305.2 | Intron | NP_851822.1 | ||||
NM_181306.2 | Intron | NP_851823.1 | ||||
NM_181307.2 | Intron | NP_851824.1 | ||||
XM_005267325.4 | Intron | XP_005267382.1 | ||||
XM_005267328.3 | Intron | XP_005267385.1 |
Set Membership: |
HapMap |