Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615194 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DHRS2 - dehydrogenase/reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318835.1 | Intron | NP_001305764.1 | ||||
NM_005794.3 | Intron | NP_005785.1 | ||||
NM_182908.4 | Intron | NP_878912.1 | ||||
XM_005267249.1 | Intron | XP_005267306.1 | ||||
XM_006720001.3 | Intron | XP_006720064.1 | ||||
XM_011536338.1 | Intron | XP_011534640.1 | ||||
XM_011536339.2 | Intron | XP_011534641.1 |
Set Membership: |
HapMap |