Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTACTAGATTTTAAAATGTGTGC[A/T]TTTTCATTTTGGAGAAGATGTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604562 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACIN1 - apoptotic chromatin condensation inducer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C14orf119 - chromosome 14 open reading frame 119 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017924.3 | Intron | NP_060394.1 | ||||
XM_017021390.1 | Intron | XP_016876879.1 |
LOC100128908 - leishmanolysin homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |