Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTCTCCCAGCTCTTATGTTGAT[A/G]TTACAACATGGTGGGATCGGGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610493 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf52 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C11orf52 - chromosome 11 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DIXDC1 - DIX domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037954.3 | Intron | NP_001033043.1 | ||||
NM_001278542.1 | Intron | NP_001265471.1 | ||||
NM_033425.4 | Intron | NP_219493.1 | ||||
XM_017018466.1 | Intron | XP_016873955.1 | ||||
XM_017018467.1 | Intron | XP_016873956.1 | ||||
XM_017018468.1 | Intron | XP_016873957.1 | ||||
XM_017018469.1 | Intron | XP_016873958.1 |
HSPB2-C11orf52 - HSPB2-C11orf52 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |