Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604153 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00471 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00471 - long intergenic non-protein coding RNA 471 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMUR1 - neuromedin U receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006056.4 | 1443 | Intron | NP_006047.3 | |||
XM_006712195.3 | 1443 | Intron | XP_006712258.1 | |||
XM_006712196.3 | 1443 | Intron | XP_006712259.1 | |||
XM_011510487.2 | 1443 | Intron | XP_011508789.1 | |||
XM_011510488.2 | 1443 | UTR 3 | XP_011508790.1 | |||
XM_011510489.2 | 1443 | Intron | XP_011508791.1 |