Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606502 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MS4A14 - membrane spanning 4-domains A14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079692.2 | 472 | UTR 5 | NP_001073160.1 | |||
NM_001261827.1 | 472 | UTR 5 | NP_001248756.1 | |||
NM_001261828.1 | 472 | UTR 5 | NP_001248757.1 | |||
NM_032597.4 | 472 | UTR 5 | NP_115986.3 |
MS4A7 - membrane spanning 4-domains A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021201.4 | 472 | Intron | NP_067024.1 | |||
NM_206938.1 | 472 | Intron | NP_996821.1 | |||
NM_206939.1 | 472 | Intron | NP_996822.1 | |||
NM_206940.1 | 472 | Intron | NP_996823.1 |
Set Membership: |
HapMap |