Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609374 MIM: 602640 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDCA5 - cell division cycle associated 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080668.3 | Intron | NP_542399.1 | ||||
XM_005273733.3 | Intron | XP_005273790.1 | ||||
XM_005273734.3 | Intron | XP_005273791.1 | ||||
XM_011544743.2 | Intron | XP_011543045.1 | ||||
XM_011544747.2 | Intron | XP_011543049.1 | ||||
XM_011544748.2 | Intron | XP_011543050.1 | ||||
XM_011544749.2 | Intron | XP_011543051.1 | ||||
XM_017017158.1 | Intron | XP_016872647.1 |
NAALADL1 - N-acetylated alpha-linked acidic dipeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |