Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609802 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYEF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MYEF2 - myelin expression factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301210.1 | Intron | NP_001288139.1 | ||||
NM_016132.4 | Intron | NP_057216.2 | ||||
XM_005254422.4 | Intron | XP_005254479.2 | ||||
XM_005254424.4 | Intron | XP_005254481.2 | ||||
XM_005254425.4 | Intron | XP_005254482.2 | ||||
XM_005254427.4 | Intron | XP_005254484.1 | ||||
XM_006720553.3 | Intron | XP_006720616.1 | ||||
XM_011521657.2 | Intron | XP_011519959.1 | ||||
XM_017022285.1 | Intron | XP_016877774.1 | ||||
XM_017022286.1 | Intron | XP_016877775.1 | ||||
XM_017022287.1 | Intron | XP_016877776.1 | ||||
XM_017022288.1 | Intron | XP_016877777.1 | ||||
XM_017022289.1 | Intron | XP_016877778.1 | ||||
XM_017022290.1 | Intron | XP_016877779.1 | ||||
XM_017022291.1 | Intron | XP_016877780.1 | ||||
XM_017022292.1 | Intron | XP_016877781.1 |
SLC24A5 - solute carrier family 24 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |