Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTGGGAAACTTCACAGAATTC[C/T]TCCTTGCTTTTTCTCAGTATTCAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608289 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF2BP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGF2BP2 - insulin like growth factor 2 mRNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007225.1 | Intron | NP_001007226.1 | ||||
NM_001291869.1 | Intron | NP_001278798.1 | ||||
NM_001291872.1 | Intron | NP_001278801.1 | ||||
NM_001291873.1 | Intron | NP_001278802.1 | ||||
NM_001291874.1 | Intron | NP_001278803.1 | ||||
NM_001291875.1 | Intron | NP_001278804.1 | ||||
NM_006548.4 | Intron | NP_006539.3 | ||||
XM_006713477.3 | Intron | XP_006713540.1 | ||||
XM_006713478.3 | Intron | XP_006713541.1 | ||||
XM_017005557.1 | Intron | XP_016861046.1 | ||||
XM_017005558.1 | Intron | XP_016861047.1 | ||||
XM_017005559.1 | Intron | XP_016861048.1 | ||||
XM_017005560.1 | Intron | XP_016861049.1 | ||||
XM_017005561.1 | Intron | XP_016861050.1 | ||||
XM_017005562.1 | Intron | XP_016861051.1 | ||||
XM_017005563.1 | Intron | XP_016861052.1 | ||||
XM_017005564.1 | Intron | XP_016861053.1 |
IGF2BP2-AS1 - IGF2BP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |