Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACAGATCCCGACATAAACATCT[T/G]CAAACTTGATGGGTTTTACGTGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603094 MIM: 611931 | ||||||||||||||||||||
Literature Links: |
B3GALNT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B3GALNT1 - beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038628.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | NP_001033717.1 | |
NM_003781.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | NP_003772.1 | |
NM_033167.2 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | NP_149357.1 | |
NM_033168.2 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | NP_149358.1 | |
NM_033169.2 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | NP_149359.1 | |
XM_005247859.4 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_005247916.1 | |
XM_005247861.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_005247918.1 | |
XM_005247862.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_005247919.1 | |
XM_005247864.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_005247921.1 | |
XM_006713803.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_006713866.1 | |
XM_006713804.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_006713867.1 | |
XM_006713805.3 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_006713868.1 | |
XM_017007402.1 | 1425 | Missense Mutation | GAA,GCA | E,A 386 | XP_016862891.1 | |
XM_017007403.1 | 1425 | Missense Mutation | GAA,GCA | E,A 386 | XP_016862892.1 | |
XM_017007404.1 | 1425 | Missense Mutation | GAA,GCA | E,A 386 | XP_016862893.1 | |
XM_017007405.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_016862894.1 | |
XM_017007406.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_016862895.1 | |
XM_017007407.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_016862896.1 | |
XM_017007408.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_016862897.1 | |
XM_017007409.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_016862898.1 | |
XM_017007410.1 | 1425 | Missense Mutation | GAA,GCA | E,A 266 | XP_016862899.1 | |
XM_017007411.1 | 1425 | UTR 3 | XP_016862900.1 | |||
XM_017007412.1 | 1425 | UTR 3 | XP_016862901.1 | |||
XM_017007413.1 | 1425 | Intron | XP_016862902.1 |
PPM1L - protein phosphatase, Mg2+/Mn2+ dependent 1L | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |