Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600293 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADCY5 - adenylate cyclase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199642.1 | Intron | NP_001186571.1 | ||||
NM_183357.2 | Intron | NP_899200.1 | ||||
XM_005247077.3 | Intron | XP_005247134.1 | ||||
XM_005247078.1 | Intron | XP_005247135.1 | ||||
XM_006713483.1 | Intron | XP_006713546.1 | ||||
XM_006713484.1 | Intron | XP_006713547.1 | ||||
XM_011512359.2 | Intron | XP_011510661.1 | ||||
XM_011512360.2 | Intron | XP_011510662.1 | ||||
XM_011512361.1 | Intron | XP_011510663.1 | ||||
XM_017005638.1 | Intron | XP_016861127.1 | ||||
XM_017005639.1 | Intron | XP_016861128.1 | ||||
XM_017005640.1 | Intron | XP_016861129.1 |