Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC191 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC191 - coiled-coil domain containing 191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QTRT2 - queuine tRNA-ribosyltransferase accessory subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256835.1 | Intron | NP_001243764.1 | ||||
NM_001256836.1 | Intron | NP_001243765.1 | ||||
NM_001256837.1 | Intron | NP_001243766.1 | ||||
NM_024638.3 | Intron | NP_078914.1 | ||||
XM_005247769.3 | Intron | XP_005247826.1 | ||||
XM_005247770.3 | Intron | XP_005247827.1 | ||||
XM_006713744.3 | Intron | XP_006713807.1 | ||||
XM_011513137.2 | Intron | XP_011511439.1 | ||||
XM_017007182.1 | Intron | XP_016862671.1 | ||||
XM_017007183.1 | Intron | XP_016862672.1 | ||||
XM_017007184.1 | Intron | XP_016862673.1 |
Set Membership: |
HapMap |