Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615864 | ||||||||||||||||||||
Literature Links: |
CEP97 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP97 - centrosomal protein 97 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303401.1 | 64 | UTR 5 | NP_001290330.1 | |||
NM_024548.3 | 64 | UTR 5 | NP_078824.2 | |||
XM_006713743.3 | 64 | UTR 5 | XP_006713806.1 | |||
XM_011513127.1 | 64 | Intron | XP_011511429.1 | |||
XM_017007178.1 | 64 | UTR 5 | XP_016862667.1 |