Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603581 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCDH9 - protocadherin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318372.1 | Intron | NP_001305301.1 | ||||
NM_001318373.1 | Intron | NP_001305302.1 | ||||
NM_001318374.1 | Intron | NP_001305303.1 | ||||
NM_020403.4 | Intron | NP_065136.1 | ||||
NM_203487.2 | Intron | NP_982354.1 | ||||
XM_005266408.3 | Intron | XP_005266465.1 | ||||
XM_011535099.2 | Intron | XP_011533401.1 | ||||
XM_017020619.1 | Intron | XP_016876108.1 | ||||
XM_017020620.1 | Intron | XP_016876109.1 | ||||
XM_017020621.1 | Intron | XP_016876110.1 |
Set Membership: |
HapMap |