Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601647 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R5E PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP2R5E - protein phosphatase 2 regulatory subunit B'epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282179.1 | Intron | NP_001269108.1 | ||||
NM_001282180.1 | Intron | NP_001269109.1 | ||||
NM_001282181.1 | Intron | NP_001269110.1 | ||||
NM_001282182.1 | Intron | NP_001269111.1 | ||||
NM_006246.3 | Intron | NP_006237.1 |
Set Membership: |
HapMap |