Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf105 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C14orf105 - chromosome 14 open reading frame 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283056.1 | Intron | NP_001269985.1 | ||||
NM_001283057.1 | Intron | NP_001269986.1 | ||||
NM_001283058.1 | Intron | NP_001269987.1 | ||||
NM_001283059.1 | Intron | NP_001269988.1 | ||||
NM_001283060.1 | Intron | NP_001269989.1 | ||||
NM_018168.3 | Intron | NP_060638.2 | ||||
XM_005267806.1 | Intron | XP_005267863.1 | ||||
XM_005267810.3 | Intron | XP_005267867.1 | ||||
XM_005267811.3 | Intron | XP_005267868.1 | ||||
XM_005267813.1 | Intron | XP_005267870.1 | ||||
XM_005267814.1 | Intron | XP_005267871.1 | ||||
XM_006720188.2 | Intron | XP_006720251.1 | ||||
XM_006720189.2 | Intron | XP_006720252.1 |