Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCAGGAGCTTTCGCTCTTACGG[C/G]CTCAAGAGATATCATCTAACCAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604807 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLRT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLRT2 - fibronectin leucine rich transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013231.4 | Intron | NP_037363.1 | ||||
XM_005267490.3 | Intron | XP_005267547.1 | ||||
XM_011536611.2 | Intron | XP_011534913.1 | ||||
XM_017021129.1 | Intron | XP_016876618.1 | ||||
XM_017021130.1 | Intron | XP_016876619.1 | ||||
XM_017021131.1 | Intron | XP_016876620.1 | ||||
XM_017021132.1 | Intron | XP_016876621.1 | ||||
XM_017021133.1 | Intron | XP_016876622.1 | ||||
XM_017021134.1 | Intron | XP_016876623.1 | ||||
XM_017021135.1 | Intron | XP_016876624.1 |
LOC100506731 - uncharacterized LOC100506731 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |