Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAATGCCTGATGAGAGCAAGG[C/T]GCAAGATCTGTGGAGCTCAAGGGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611121 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLMN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLMN - calmin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024734.3 | 9667 | UTR 3 | NP_079010.2 | |||
XM_011537158.1 | 9667 | UTR 3 | XP_011535460.1 | |||
XM_011537159.2 | 9667 | Intron | XP_011535461.1 | |||
XM_017021646.1 | 9667 | UTR 3 | XP_016877135.1 | |||
XM_017021647.1 | 9667 | Intron | XP_016877136.1 |
DICER1-AS1 - DICER1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |