Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602462 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C4orf50 - chromosome 4 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011513603.2 | 14226 | UTR 3 | XP_011511905.1 | |||
XM_011513604.2 | 14226 | UTR 3 | XP_011511906.1 | |||
XM_017008891.1 | 14226 | UTR 3 | XP_016864380.1 | |||
XM_017008892.1 | 14226 | UTR 3 | XP_016864381.1 | |||
XM_017008893.1 | 14226 | UTR 3 | XP_016864382.1 | |||
XM_017008894.1 | 14226 | Intron | XP_016864383.1 |
CRMP1 - collapsin response mediator protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |