Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607573 | ||||||||||||||||||||
Literature Links: |
MAP1S PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP1S - microtubule associated protein 1S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308363.1 | 579 | Silent Mutation | AGA,CGA | R,R 37 | NP_001295292.1 | |
NM_018174.5 | 579 | Silent Mutation | AGA,CGA | R,R 63 | NP_060644.4 | |
XM_017026930.1 | 579 | Silent Mutation | AGA,CGA | R,R 63 | XP_016882419.1 | |
XM_017026931.1 | 579 | Missense Mutation | AGA,CGA | R,R 37 | XP_016882420.1 |