Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138253 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN2A - glutamate ionotropic receptor NMDA type subunit 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000833.4 | Intron | NP_000824.1 | ||||
NM_001134407.2 | Intron | NP_001127879.1 | ||||
NM_001134408.2 | Intron | NP_001127880.1 | ||||
XM_011522458.2 | Intron | XP_011520760.1 | ||||
XM_011522461.2 | Intron | XP_011520763.1 | ||||
XM_017023172.1 | Intron | XP_016878661.1 | ||||
XM_017023173.1 | Intron | XP_016878662.1 |
IMPDH1P11 - inosine monophosphate dehydrogenase 1 pseudogene 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |