Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611761 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LMF1 - lipase maturation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022773.2 | Intron | NP_073610.2 | ||||
XM_006720928.2 | Intron | XP_006720991.1 | ||||
XM_006720929.3 | Intron | XP_006720992.1 | ||||
XM_011522614.2 | Intron | XP_011520916.1 | ||||
XM_011522615.1 | Intron | XP_011520917.1 | ||||
XM_011522617.2 | Intron | XP_011520919.1 | ||||
XM_011522618.2 | Intron | XP_011520920.1 | ||||
XM_017023575.1 | Intron | XP_016879064.1 | ||||
XM_017023576.1 | Intron | XP_016879065.1 | ||||
XM_017023577.1 | Intron | XP_016879066.1 | ||||
XM_017023578.1 | Intron | XP_016879067.1 | ||||
XM_017023579.1 | Intron | XP_016879068.1 | ||||
XM_017023580.1 | Intron | XP_016879069.1 | ||||
XM_017023581.1 | Intron | XP_016879070.1 |